Assay Details
| Gene Symbol: ABRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135960.2 | NP_001129432.1 | 1 - 2 | 731 | 74 | |
| GenBank mRNA | AK347188.1 | - | 1 - 2 | 83 | 74 | |
| AY609571.1 | - | 1 - 2 | 716 | 74 | ||
| DQ536197.1 | - | 1 - 2 | 732 | 74 | ||
More Information
Set Membership: |
|

ABRA
Genomic Map