Assay Details
| Gene Symbol: PFKFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143721.1 | NP_001137193.1 | 6 - 7 | 749 | 60 | |
| XM_005673663.3 | XP_005673720.1 | 861 | 60 | |||
| XM_005673664.3 | XP_005673721.1 | 676 | 60 | |||
| XM_013986195.2 | XP_013841649.1 | 854 | 60 | |||
| XM_021079416.1 | XP_020935075.1 | 1210 | 60 | |||
| XM_021079417.1 | XP_020935076.1 | 558 | 60 | |||
| XM_021079418.1 | XP_020935077.1 | 1642 | 60 | |||
| XM_021079419.1 | XP_020935078.1 | 781 | 60 | |||
| XM_021079420.1 | XP_020935079.1 | 676 | 60 | |||
| XM_021079421.1 | XP_020935080.1 | 573 | 60 | |||
| XM_021079422.1 | XP_020935081.1 | 701 | 60 | |||
| GenBank mRNA | AK347129.1 | - | 6 - 7 | 541 | 60 | |
| AK394635.1 | - | 6 - 7 | 510 | 60 | ||
| FJ436397.1 | - | 6 - 7 | 749 | 60 | ||
More Information
Set Membership: |
|

PFKFB1
Genomic Map