Assay Details
| Gene Symbol: ID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123130.1 | NP_001116602.1 | 1 | 50 | 53 | |
| XM_005665612.3 | XP_005665669.2 | 471 | 53 | |||
| GenBank mRNA | DQ917620.1 | - | 1 | 50 | 53 | |
More Information
Set Membership: |
|

ID4
Genomic Map