Assay Details
| Gene Symbol: MEST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128471.1 | NP_001121943.1 | 11 | 1151 | 70 | |
| XM_005673159.3 | XP_005673216.1 | 1454 | 70 | |||
| XM_005673160.3 | XP_005673217.1 | 1265 | 70 | |||
| XM_005673161.3 | XP_005673218.1 | 1291 | 70 | |||
| XM_021078439.1 | XP_020934098.1 | 1165 | 70 | |||
| XM_021078440.1 | XP_020934099.1 | 1165 | 70 | |||
| XM_021078441.1 | XP_020934100.1 | 2027 | 70 | |||
| GenBank mRNA | EF546431.1 | - | 11 | 1151 | 70 | |
| EF619473.1 | - | 12 | 1159 | 70 | ||
| HM126537.1 | - | 12 | 1159 | 70 | ||
More Information
Set Membership: |
|

MEST
Genomic Map