Assay Details
| Gene Symbol: FKBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128485.1 | NP_001121957.1 | 1 | 650 | 113 | |
| XM_005665825.3 | XP_005665882.1 | 947 | 113 | |||
| XM_013977654.2 | XP_013833108.1 | 762 | 113 | |||
| XM_021098042.1 | XP_020953701.1 | 1007 | 113 | |||
| GenBank mRNA | AK345965.1 | - | 2 | 923 | 113 | |
| AK352399.1 | - | 2 | 977 | 113 | ||
| EU282364.1 | - | 1 | 745 | 113 | ||
More Information
Set Membership: |
|

FKBPL
Genomic Map