Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129970.1 | NP_001123442.1 | 3 | 1146 | 95 | |
| GenBank mRNA | AK391860.1 | - | 3 | 1160 | 95 | |
| AK399566.1 | - | 3 | 1189 | 95 | ||
| AY864613.1 | - | 3 | 1146 | 95 | ||
More Information
Set Membership: |
|

CYCS
Genomic Map