Assay Details
| Gene Symbol: KLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134350.2 | NP_001127822.2 | 2 | 804 | 80 | |
| GenBank mRNA | AK399764.1 | - | 2 | 795 | 80 | |
| EU669072.2 | - | 2 | 804 | 80 | ||
More Information
Set Membership: |
|

KLF1
Genomic Map