Assay Details
| Gene Symbol: FOXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135959.1 | NP_001129431.1 | 2 | 1945 | 112 | |
| XM_021084213.1 | XP_020939872.1 | 2324 | 112 | |||
| XM_021084221.1 | XP_020939880.1 | 2215 | 112 | |||
| XM_021084231.1 | XP_020939890.1 | 2176 | 112 | |||
| GenBank mRNA | AB433530.1 | - | 2 | 1945 | 112 | |
| AK395232.1 | - | 1 | 541 | 112 | ||
| DQ243691.2 | - | 1 | 73 | 112 | ||
More Information
Set Membership: |
|

FOXO3
Genomic Map