Assay Details
| Gene Symbol: ERO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137627.1 | NP_001131099.1 | 12 | 955 | 98 | |
| GenBank mRNA | EU552934.1 | - | 12 | 955 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ERO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137627.1 | NP_001131099.1 | 12 | 955 | 98 | |
| GenBank mRNA | EU552934.1 | - | 12 | 955 | 98 | |
Set Membership: |
|