Assay Details
| Gene Symbol: POT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137635.2 | NP_001131107.1 | 6 | 751 | 93 | |
| XM_005673197.3 | XP_005673254.1 | 1697 | 93 | |||
| XM_005673198.3 | XP_005673255.1 | 1734 | 93 | |||
| XM_013985634.2 | XP_013841088.1 | 1637 | 93 | |||
| XM_021078450.1 | XP_020934109.1 | 1575 | 93 | |||
| GenBank mRNA | FJ167678.1 | - | 6 | 754 | 93 | |
More Information
Set Membership: |
|

POT1
Genomic Map