Assay Details
| Gene Symbol: TEAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142666.1 | NP_001136138.1 | 11 | 1350 | 75 | |
| GenBank mRNA | AK240248.1 | - | 13 | 1541 | 75 | |
| AK347910.1 | - | 12 | 1616 | 75 | ||
| EU286538.1 | - | 11 | 1350 | 75 | ||
| EU286539.1 | - | 10 | 1221 | 75 | ||
More Information
Set Membership: |
|

TEAD4
Genomic Map