Assay Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142832.1 | NP_001136304.1 | 7 | 953 | 102 | |
| XM_021076525.1 | XP_020932184.1 | 1173 | 102 | |||
| GenBank mRNA | EU725862.1 | - | 7 | 953 | 102 | |
More Information
Set Membership: |
|

MAT2B
Genomic Map