Assay Details
| Gene Symbol: XBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142836.1 | NP_001136308.1 | 5 | 1445 | 93 | |
| NM_001271738.1 | NP_001258667.1 | 6 | 1419 | 93 | ||
| GenBank mRNA | AK233029.1 | - | 5 | 1445 | 93 | |
| AK392484.1 | - | 5 | 1446 | 93 | ||
| FJ213449.1 | - | 5 | 1445 | 93 | ||
More Information
Set Membership: |
|

XBP1
Genomic Map