Assay Details
| Gene Symbol: GZMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143693.1 | NP_001137165.1 | 1 | 154 | 80 | |
| GenBank mRNA | AK238642.1 | - | 2 | 435 | 80 | |
| AK351625.1 | - | 1 | 152 | 80 | ||
| FJ439673.1 | - | 1 | 154 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143693.1 transcripts.
Set Membership: |
|

GZMH
Genomic Map