Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143696.1 | NP_001137168.1 | 7 | 947 | 91 | |
| XM_021071996.1 | XP_020927655.1 | 1413 | 91 | |||
| GenBank mRNA | AK400162.1 | - | 7 | 1575 | 91 | |
| FJ436380.1 | - | 7 | 947 | 91 | ||
More Information
Set Membership: |
|

PTEN
Genomic Map