Assay Details
| Gene Symbol: STARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143726.1 | NP_001137198.1 | 6 | 555 | 106 | |
| XM_021077159.1 | XP_020932818.1 | 702 | 106 | |||
| GenBank mRNA | FJ436387.1 | - | 6 | 555 | 106 | |
More Information
Set Membership: |
|

STARD4
Genomic Map