Assay Details
| Gene Symbol: VGLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143929.1 | NP_001137401.1 | 3 | 548 | 100 | |
| XM_005657128.3 | XP_005657185.1 | 932 | 100 | |||
| XM_013990172.2 | XP_013845626.1 | 914 | 100 | |||
| XM_013990173.2 | XP_013845627.1 | 896 | 100 | |||
| GenBank mRNA | FJ481172.1 | - | 3 | 548 | 100 | |
| FJ481173.1 | - | 3 | 380 | 100 | ||
More Information
Set Membership: |
|

VGLL3
Genomic Map