Assay Details
| Gene Symbol: TPRKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144846.1 | NP_001138318.1 | 3 | 246 | 81 | |
| XM_005662459.2 | XP_005662516.1 | 1657 | 81 | |||
| XM_005662460.2 | XP_005662517.1 | 1726 | 81 | |||
| XM_005662461.2 | XP_005662518.1 | 1613 | 81 | |||
| XM_021085603.1 | XP_020941262.1 | 1783 | 81 | |||
| XM_021085604.1 | XP_020941263.1 | 1758 | 81 | |||
| GenBank mRNA | AK347171.1 | - | 4 | 420 | 81 | |
| AY610411.1 | - | 3 | 245 | 81 | ||
| FJ487631.1 | - | 3 | 246 | 81 | ||
More Information
Set Membership: |
|

TPRKB
Genomic Map