Assay Details
| Gene Symbol: PSMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144901.1 | NP_001138373.1 | 3 | 249 | 76 | |
| XM_021088747.1 | XP_020944406.1 | 169 | 76 | |||
| XM_021088748.1 | XP_020944407.1 | 178 | 76 | |||
| GenBank mRNA | AK237173.1 | - | 3 | 249 | 76 | |
| AK392939.1 | - | 3 | 255 | 76 | ||
| BW981249.1 | - | 3 | 249 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021088748.1 transcripts.
Set Membership: |
|

PSMA5
Genomic Map