Assay Details
| Gene Symbol: RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145223.1 | NP_001138695.1 | 4 | 647 | 95 | |
| XM_021090967.1 | XP_020946626.1 | 872 | 95 | |||
| GenBank mRNA | AK232352.1 | - | 4 | 698 | 95 | |
| AK394203.1 | - | 4 | 694 | 95 | ||
| EF208120.1 | - | 4 | 647 | 95 | ||
More Information
Set Membership: |
|

RBP5
Genomic Map