Assay Details
| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145379.2 | NP_001138851.1 | 5 | 1350 | 105 | |
| GenBank mRNA | AK233123.1 | - | 5 | 1183 | 105 | |
| AK239636.1 | - | 4 | 1078 | 105 | ||
| AK345430.1 | - | 4 | 1040 | 105 | ||
| AK348704.1 | - | 5 | 1117 | 105 | ||
| FJ494915.1 | - | 5 | 1351 | 105 | ||
More Information
Set Membership: |
|

ICAM3
Genomic Map