Assay Details
| Gene Symbol: ATP6V1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145380.1 | NP_001138852.1 | 3 | 255 | 97 | |
| XM_021097986.1 | XP_020953645.1 | 738 | 97 | |||
| XM_021097987.1 | XP_020953646.1 | 518 | 97 | |||
| XM_021097988.1 | XP_020953647.1 | 386 | 97 | |||
| GenBank mRNA | AK392664.1 | - | 3 | 475 | 97 | |
| AK400040.1 | - | 3 | 367 | 97 | ||
| EU282339.1 | - | 3 | 494 | 97 | ||
More Information
Set Membership: |
|

ATP6V1G2
Genomic Map