Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159311.1 | NP_001152783.1 | 14 | 2548 | 81 | |
| GenBank mRNA | FJ597911.1 | - | 14 | 2548 | 81 | |
More Information
Set Membership: |
|

SPAG5
Genomic Map