Assay Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159312.1 | NP_001152784.1 | 8 | 1255 | 95 | |
| GenBank mRNA | AK238409.1 | - | 4 | 603 | 95 | |
| FJ597908.1 | - | 8 | 1255 | 95 | ||
More Information
Set Membership: |
|

SPAG6
Genomic Map