Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214314.2 | NP_999479.2 | 2 | 1624 | 86 | |
| XM_013986227.2 | XP_013841681.1 | 2106 | 86 | |||
| XM_021079513.1 | XP_020935172.1 | 2873 | 86 | |||
| GenBank mRNA | AF161717.1 | - | 3 | 2048 | 86 | |
| AF202775.1 | - | 2 | 1624 | 86 | ||
More Information
Set Membership: |
|

AR
Genomic Map