Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129958.1 | NP_001123430.1 | 5 - 6 | 1181 | 63 | |
| GenBank mRNA | AK394174.1 | - | 5 - 6 | 1265 | 63 | |
| DQ530373.1 | - | 5 - 6 | 1181 | 63 | ||
More Information
Set Membership: |
|

SERPINC1
Genomic Map