Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164113.1 | NP_001157585.1 | 3 - 4 | 351 | 99 | |
| GenBank mRNA | AY609646.1 | - | 3 - 4 | 342 | 99 | |
| BI341962.1 | - | 3 - 4 | 351 | 99 | ||
| GU983865.1 | - | 3 - 4 | 311 | 99 | ||
More Information
Set Membership: |
|

SELENOS
Genomic Map