Assay Details
| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164509.1 | NP_001157981.1 | 1 - 2 | 107 | 67 | |
| GenBank mRNA | AK240196.1 | - | 1 - 2 | 213 | 67 | |
| EF397415.1 | - | 1 - 2 | 107 | 67 | ||
More Information
Set Membership: |
|

CRABP2
Genomic Map