Assay Details
| Gene Symbol: SLC25A19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164514.1 | NP_001157986.1 | 6 - 7 | 814 | 60 | |
| XM_021066050.1 | XP_020921709.1 | 870 | 60 | |||
| XM_021066051.1 | XP_020921710.1 | 714 | 60 | |||
| GenBank mRNA | AK236644.1 | - | 6 - 7 | 798 | 60 | |
| GQ368240.1 | - | 6 - 7 | 814 | 60 | ||
More Information
Set Membership: |
|

SLC25A19
Genomic Map