Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164515.1 | NP_001157987.1 | 1 - 2 | 99 | 64 | |
| GenBank mRNA | AK391248.1 | - | 1 - 2 | 149 | 64 | |
| AK399557.1 | - | 1 - 2 | 149 | 64 | ||
| GQ338308.1 | - | 1 - 2 | 99 | 64 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map