Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164515.1 | NP_001157987.1 | 2 - 3 | 217 | 66 | |
| GenBank mRNA | AK391248.1 | - | 2 - 3 | 267 | 66 | |
| FJ156752.1 | - | 2 - 3 | 263 | 66 | ||
| GQ338308.1 | - | 2 - 3 | 217 | 66 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map