Assay Details
| Gene Symbol: SMNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166309.1 | NP_001159781.1 | 4 - 5 | 617 | 64 | |
| XM_005671451.3 | XP_005671508.1 | 795 | 64 | |||
| GenBank mRNA | AK232698.1 | - | 4 - 5 | 593 | 64 | |
| AY609548.1 | - | 4 - 5 | 570 | 64 | ||
| EU571478.1 | - | 4 - 5 | 617 | 64 | ||
More Information
Set Membership: |
|

SMNDC1
Genomic Map