Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166313.1 | NP_001159785.1 | 1 - 2 | 114 | 68 | |
| XM_005661543.3 | XP_005661600.1 | 676 | 68 | |||
| XM_005661544.3 | XP_005661601.1 | 343 | 68 | |||
| XM_013994968.2 | XP_013850422.1 | 676 | 68 | |||
| XM_013994969.2 | XP_013850423.1 | 676 | 68 | |||
| XM_013994970.2 | XP_013850424.1 | 676 | 68 | |||
| XM_013994971.2 | XP_013850425.1 | 676 | 68 | |||
| XM_013994972.2 | XP_013850426.1 | 676 | 68 | |||
| XM_021077281.1 | XP_020932940.1 | 676 | 68 | |||
| XM_021077311.1 | XP_020932970.1 | 676 | 68 | |||
| GenBank mRNA | AB499892.1 | - | 1 - 2 | 114 | 68 | |
More Information
Set Membership: |
|

CCNH
Genomic Map