Assay Details
| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128450.1 | NP_001121922.1 | 4 - 5 | 522 | 72 | |
| GenBank mRNA | EU282371.1 | - | 5 - 6 | 1195 | 72 | |
More Information
Set Membership: |
|

STK19
Genomic Map