Assay Details
| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128473.1 | NP_001121945.1 | 10 - 11 | 1831 | 96 | |
| XM_021088503.1 | XP_020944162.1 | 96 | ||||
| XM_021088504.1 | XP_020944163.1 | 96 | ||||
| GenBank mRNA | AK234627.1 | - | 10 - 11 | 96 | ||
| AK239047.1 | - | 10 - 11 | 96 | |||
| AK391444.1 | - | 10 - 11 | 96 | |||
| AK393077.1 | - | 6 - 7 | 96 | |||
| EU259887.1 | - | 10 - 11 | 1831 | 96 | ||
| KF977840.1 | - | 9 - 10 | 96 | |||
More Information
Set Membership: |
|

GBP1
Genomic Map