Assay Details
| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128473.1 | NP_001121945.1 | 7 - 8 | 1311 | 108 | |
| XM_021088503.1 | XP_020944162.1 | 1343 | 108 | |||
| XM_021088504.1 | XP_020944163.1 | 1330 | 108 | |||
| GenBank mRNA | AK234627.1 | - | 7 - 8 | 1315 | 108 | |
| AK239047.1 | - | 7 - 8 | 1233 | 108 | ||
| AK391444.1 | - | 7 - 8 | 1286 | 108 | ||
| AK393077.1 | - | 3 - 4 | 1137 | 108 | ||
| EU259887.1 | - | 7 - 8 | 1311 | 108 | ||
| KF977840.1 | - | 6 - 7 | 1142 | 108 | ||
More Information
Set Membership: |
|

GBP1
Genomic Map