Assay Details
| Gene Symbol: GBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128474.1 | NP_001121946.1 | 10 - 11 | 1928 | 75 | |
| GenBank mRNA | AK239977.1 | - | 6 - 7 | 1091 | 75 | |
| AK344864.1 | - | 7 - 8 | 1280 | 75 | ||
| AK345662.1 | - | 9 - 10 | 1715 | 75 | ||
| AK393079.1 | - | 10 - 11 | 1920 | 75 | ||
| EU259888.1 | - | 10 - 11 | 1928 | 75 | ||
More Information
Set Membership: |
|

GBP2
Genomic Map