Assay Details
| Gene Symbol: GBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128474.1 | NP_001121946.1 | 1 - 2 | 247 | 120 | |
| GenBank mRNA | AK393079.1 | - | 1 - 2 | 239 | 120 | |
| EU259888.1 | - | 1 - 2 | 247 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001128474.1 transcripts.
Set Membership: |
|

GBP2
Genomic Map