Assay Details
| Gene Symbol: GBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128474.1 | NP_001121946.1 | 9 - 10 | 1738 | 106 | |
| GenBank mRNA | AK238343.1 | - | 6 - 7 | 1135 | 106 | |
| AK239977.1 | - | 5 - 6 | 901 | 106 | ||
| AK344864.1 | - | 6 - 7 | 1090 | 106 | ||
| AK345662.1 | - | 8 - 9 | 1525 | 106 | ||
| AK393079.1 | - | 9 - 10 | 1730 | 106 | ||
| EU259888.1 | - | 9 - 10 | 1738 | 106 | ||
More Information
Set Membership: |
|

GBP2
Genomic Map