Assay Details
| Gene Symbol: PROX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128490.1 | NP_001121962.1 | 1 - 2 | 1724 | 96 | |
| XM_005656752.3 | XP_005656809.1 | 2223 | 96 | |||
| XM_005656753.3 | XP_005656810.1 | 1921 | 96 | |||
| XM_005656754.3 | XP_005656811.1 | 1982 | 96 | |||
| XM_005656755.3 | XP_005656812.1 | 1883 | 96 | |||
| XM_005656756.3 | XP_005656813.1 | 3127 | 96 | |||
| XM_005656757.3 | XP_005656814.1 | 2530 | 96 | |||
| XM_013989660.2 | XP_013845114.1 | 1878 | 96 | |||
| XM_013989661.2 | XP_013845115.1 | 1872 | 96 | |||
| XM_013989662.2 | XP_013845116.1 | 1905 | 96 | |||
| XM_021101942.1 | XP_020957601.1 | 2571 | 96 | |||
| XM_021101943.1 | XP_020957602.1 | 2305 | 96 | |||
| XM_021101944.1 | XP_020957603.1 | 4132 | 96 | |||
| XM_021101945.1 | XP_020957604.1 | 1910 | 96 | |||
| GenBank mRNA | AK394044.1 | - | 1 - 2 | 545 | 96 | |
| EF486324.1 | - | 1 - 2 | 1724 | 96 | ||
More Information
Set Membership: |
|

PROX1
Genomic Map