Assay Details
| Gene Symbol: LGALS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142841.1 | NP_001136313.1 | 3 - 4 | 344 | 71 | |
| GenBank mRNA | FJ424703.1 | - | 3 - 4 | 344 | 71 | |
More Information
Set Membership: |
|

LGALS13
Genomic Map