Assay Details
| Gene Symbol: PNLIPRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142897.1 | NP_001136369.1 | 4 - 5 | 481 | 58 | |
| GenBank mRNA | FJ458419.1 | - | 4 - 5 | 481 | 58 | |
More Information
Set Membership: |
|

PNLIPRP1
Genomic Map