Assay Details
| Gene Symbol: CPNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143703.1 | NP_001137175.1 | 13 - 14 | 1202 | 79 | |
| GenBank mRNA | FJ436394.1 | - | 13 - 14 | 1202 | 79 | |
More Information
Set Membership: |
|

CPNE1
Genomic Map