Assay Details
| Gene Symbol: NME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143720.1 | NP_001137192.1 | 3 - 4 | 328 | 71 | |
| XM_021085597.1 | XP_020941256.1 | 976 | 71 | |||
| XM_021085598.1 | XP_020941257.1 | 993 | 71 | |||
| XM_021085599.1 | XP_020941258.1 | 412 | 71 | |||
| XM_021085600.1 | XP_020941259.1 | 985 | 71 | |||
| GenBank mRNA | FJ436396.1 | - | 3 - 4 | 328 | 71 | |
More Information
Set Membership: |
|

NME4
Genomic Map