Assay Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143723.1 | NP_001137195.1 | 5 - 6 | 753 | 101 | |
| XM_005661427.3 | XP_005661484.2 | 2557 | 101 | |||
| XM_005661428.3 | XP_005661485.1 | 796 | 101 | |||
| GenBank mRNA | FJ436381.1 | - | 5 - 6 | 753 | 101 | |
| FJ436382.1 | - | 3 - 4 | 415 | 101 | ||
| FJ436383.1 | - | 4 - 5 | 612 | 101 | ||
More Information
Set Membership: |
|

PLPP2
Genomic Map