Assay Details
| Gene Symbol: RBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145222.2 | NP_001138694.1 | 3 - 4 | 407 | 66 | |
| GenBank mRNA | AY609848.1 | - | 3 - 4 | 414 | 66 | |
| EF208119.1 | - | 3 - 4 | 408 | 66 | ||
More Information
Set Membership: |
|

RBP7
Genomic Map