Assay Details
| Gene Symbol: LOC396566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161635.1 | NP_001155107.1 | 1 | 253 | 75 | |
| XM_005666863.2 | XP_005666920.2 | 601 | 75 | |||
| Gene Symbol: CLDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ873109.1 | - | 1 | 253 | 75 | |
More Information
Set Membership: |
|

LOC396566
Genomic Map