Assay Details
| Gene Symbol: MAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161639.1 | NP_001155111.1 | 1 - 2 | 255 | 66 | |
| GenBank mRNA | FJ603023.1 | - | 1 - 2 | 255 | 66 | |
More Information
Set Membership: |
|

MAL2
Genomic Map