Assay Details
| Gene Symbol: MAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161639.1 | NP_001155111.1 | 2 - 3 | 430 | 102 | |
| XM_005662876.3 | XP_005662933.1 | 408 | 102 | |||
| GenBank mRNA | FJ603023.1 | - | 2 - 3 | 430 | 102 | |
More Information
Set Membership: |
|

MAL2
Genomic Map