Assay Details
| Gene Symbol: MYRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161754.1 | NP_001155226.1 | 4 - 5 | 579 | 64 | |
| XM_021068254.1 | XP_020923913.1 | 962 | 64 | |||
| XM_021068255.1 | XP_020923914.1 | 278 | 64 | |||
| GenBank mRNA | FJ589745.1 | - | 4 - 5 | 579 | 64 | |
More Information
Set Membership: |
|

MYRIP
Genomic Map